Search
Search
| Gene Symbol: Zbtb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213564.3 | NP_998729.1 | 2 | 1379 | 65 | |
| XM_006256121.2 | XP_006256183.1 | 2 | 1367 | 65 | ||
| XM_006256122.2 | XP_006256184.1 | 2 | 1312 | 65 | ||
| GenBank mRNA | BC100649.1 | - | 1327 | 7932768 | 65 | |
Set Membership: |
|