Search
Search

Cntnap5b| Gene Symbol: Cntnap5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047873.1 | NP_001041338.1 | 5 - 6 | 739 | 73 | |
| XM_008769451.1 | XP_008767673.1 | 5 - 6 | 3488 | 73 | ||
| XM_008769452.1 | XP_008767674.1 | 5 - 6 | 1172 | 73 | ||
Set Membership: |
|