Search
Search

LOC498231| Gene Symbol: LOC498231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047925.1 | NP_001041390.1 | 1 - 2 | 80 | 86 | |
| GenBank mRNA | AY539895.1 | - | 1 - 2 | 80 | 86 | |
Set Membership: |
|