Search
Search

LOC499530| Gene Symbol: LOC499530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047941.1 | NP_001041406.1 | 2 - 3 | 247 | 79 | |
| XM_008760739.1 | XP_008758961.1 | 2 - 3 | 247 | 79 | ||
| GenBank mRNA | AY539940.1 | - | 2 - 3 | 247 | 79 | |
Set Membership: |
|