Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03021105_gH
          See other RT1-A2 GE Assays ›
          Gene Symbol:
          RT1-A2
          Gene Name:
          RT1 class Ia, locus A2
          Gene Aliases:
          RT1.A2(N), RT1A-2
          Chromosome Location:
          Chr.20: 5375011 - 5378536 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03021105_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RT1-A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008829.1 NP_001008829.1 1 - 2 69 147
          GenBank mRNA BC088465.1 - 1 - 2 85 7127909 147
          FQ233418.1 - 1 - 2 95 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001802.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RT1-A2

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P296155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P296155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P296155 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P296155 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P184753 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P296155 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828424 - -
          Affymetrix Rat Gene 1.0 ST Array 10828422 - -
          Affymetrix Rat Gene 1.0 ST Array 10828427 - -
          Affymetrix Rat Gene 1.0 ST Array 10828425 - -
          Affymetrix Rat Gene 1.0 ST Array 10828423 - -
          Affymetrix Rat Gene 1.0 ST Array 10828421 - -
          Affymetrix Rat Gene 1.1 ST Array 10828417 - -
          Rat Genome 230 2.0 Array 1388203_PM_x_at - -
          Rat Genome 230 2.0 Array 1380436_PM_at - -
          Rat Genome 230 2.0 Array 1380436_at - -
          Rat Genome 230 2.0 Array 1370428_x_at - -
          Rat Genome 230 2.0 Array 1371267_at - -
          Rat Genome 230 2.0 Array 1388203_x_at - -
          Rat Genome 230 2.0 Array 1370428_PM_x_at - -
          Rat Genome 230 2.0 Array 1397859_PM_x_at - -
          Rat Genome 230 2.0 Array 1371267_PM_at - -
          Rat Genome 230 2.0 Array 1397859_x_at - -
          Affymetrix HT Rat Focus Array 1380436_PM_at - -
          Affymetrix HT Rat Focus Array 1380436_at - -
          Affymetrix Rat Clariom S Assay PSR2000014550.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014554.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014547.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014544.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014553.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014542.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014549.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014546.rn.1 TC2000001802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014552.rn.1 TC2000001802.rn.1 RTA TC
          Rat Expression Set 230 1371267_at - -
          Rat Expression Set 230 1388203_PM_x_at - -
          Rat Expression Set 230 1388203_x_at - -
          Rat Expression Set 230 1370428_x_at - -
          Rat Expression Set 230 1370428_PM_x_at - -
          Rat Expression Set 230 1371267_PM_at - -
          Rat Expression Set 230 1397859_x_at - -
          Rat Expression Set 230 1397859_PM_x_at - -
          Rat Expression Set 230 1380436_PM_at - -
          Rat Expression Set 230 1380436_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828417 - -
          Rat Genome U34 Set M64795_f_at - -
          Rat Genome U34 Set AF074609mRNA_f_at - -
          Rat Genome U34 Set M64795_i_at - -
          Rat Genome U34 Set rc_AI103500_i_at - -
          Rat Genome U34 Set rc_AI103500_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349776 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline