Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03035911_s1
          See other TSKU GE Assays ›
          Gene Symbol:
          Tsku
          Gene Name:
          tsukushi, small leucine rich proteoglycan
          Gene Aliases:
          Eiih
          Chromosome Location:
          Chr.1: 163317281 - 163328687 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03035911_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsku
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009965.2 NP_001009965.1 2 1043 117
          XM_006229759.2 XP_006229821.1 2 1055 117
          XM_006229760.2 XP_006229822.1 2 1936 117
          XM_006229761.2 XP_006229823.1 2 1238 117
          GenBank mRNA AY533242.1 - 2 964 117
          BC100644.1 - 2 1043 7930712 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsku

          Gene Ontology Categories:

          Function(s)

          regulation of gene expression
          corpus callosum morphogenesis
          lateral ventricle development
          anterior commissure morphogenesis
          negative regulation of Wnt signaling pathway
          camera-type eye development
          ciliary body morphogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098979 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098979 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098979 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P438346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723823 - -
          Affymetrix Rat Gene 1.0 ST Array 10723824 - -
          Affymetrix Rat Gene 1.1 ST Array 10723822 - -
          Rat Genome 230 2.0 Array 1383315_PM_at - -
          Rat Genome 230 2.0 Array 1383315_at - -
          Affymetrix HT Rat Focus Array 1383315_PM_at - -
          Affymetrix HT Rat Focus Array 1383315_at - -
          Affymetrix Rat Clariom S Assay PSR0100048370.rn.1 TC0100006092.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048369.rn.1 TC0100006092.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048374.rn.1 TC0100006092.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048375.rn.1 TC0100006092.rn.1 RTA TC
          Rat Expression Set 230 1383315_PM_at - -
          Rat Expression Set 230 1383315_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723822 - -
          Rat Genome U34 Set rc_AA925167_at - -
          Rat Genome U34 Set rc_AI176828_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21788042 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376612 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline