Search
Search

LOC302576| Gene Symbol: LOC302576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013959.1 | NP_001013981.1 | 1 | 1452 | 165 | |
| GenBank mRNA | BC083583.1 | - | 1 | 1452 | 7131068 | 165 |
Set Membership: |
|