Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03036301_s1
          See other SERTAD2 GE Assays ›
          Gene Symbol:
          Sertad2
          Gene Name:
          SERTA domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 104786645 - 104890022 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03036301_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sertad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024903.1 NP_001020074.2 2 1047 151
          XM_006251526.2 XP_006251588.1 3 1310 151
          XM_006251527.2 XP_006251589.1 2 2432 151
          GenBank mRNA BC098047.1 - 2 1047 7457109 151

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sertad2

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell growth
          positive regulation of transcription, DNA-templated
          transcription coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39219 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61747 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774371 - -
          Affymetrix Rat Gene 1.0 ST Array 10774370 - -
          Affymetrix Rat Gene 1.1 ST Array 10774369 - -
          Rat Genome 230 2.0 Array 1375279_at - -
          Rat Genome 230 2.0 Array 1375279_PM_at - -
          Affymetrix HT Rat Focus Array 1375279_at - -
          Affymetrix HT Rat Focus Array 1375279_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400007876.rn.1 TC1400001090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007875.rn.1 TC1400001090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007880.rn.1 TC1400001090.rn.1 RTA TC
          Rat Expression Set 230 1375279_at - -
          Rat Expression Set 230 1375279_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774369 - -
          Rat Genome U34 Set rc_AI044638_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20763806 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline