Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03042246_g1
          See other ARHGAP9 GE Assays ›
          Gene Symbol:
          Arhgap9
          Gene Name:
          Rho GTPase activating protein 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 70612648 - 70620900 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03042246_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgap9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012198.1 NP_001012198.1 3 - 4 216 78
          NM_001080789.1 NP_001074258.1 4 - 5 848 78
          XM_006241464.2 XP_006241526.1 4 - 5 926 78
          XM_006241465.2 XP_006241527.1 4 - 5 926 78
          XM_006241466.2 XP_006241528.1 4 - 5 960 78
          XM_006241467.2 XP_006241529.1 4 - 5 970 78
          XM_006241468.2 XP_006241530.1 3 - 4 273 78
          GenBank mRNA BC079334.1 - 3 - 4 216 7097179 78
          BC107938.1 - 4 - 5 848 7378387 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000896.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgap9

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          positive regulation of GTPase activity
          GTPase activator activity
          phosphatidylinositol-3,4,5-trisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136447 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531324 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P652975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895870 - -
          Affymetrix Rat Gene 1.0 ST Array 10895887 - -
          Affymetrix Rat Gene 1.0 ST Array 10895872 - -
          Affymetrix Rat Gene 1.0 ST Array 10895877 - -
          Affymetrix Rat Gene 1.0 ST Array 10895868 - -
          Affymetrix Rat Gene 1.0 ST Array 10895869 - -
          Affymetrix Rat Gene 1.0 ST Array 10895882 - -
          Affymetrix Rat Gene 1.0 ST Array 10895885 - -
          Affymetrix Rat Gene 1.0 ST Array 10895881 - -
          Affymetrix Rat Gene 1.0 ST Array 10895867 - -
          Affymetrix Rat Gene 1.0 ST Array 10895875 - -
          Affymetrix Rat Gene 1.0 ST Array 10895876 - -
          Affymetrix Rat Gene 1.0 ST Array 10895886 - -
          Affymetrix Rat Gene 1.0 ST Array 10895883 - -
          Affymetrix Rat Gene 1.0 ST Array 10895884 - -
          Affymetrix Rat Gene 1.0 ST Array 10895874 - -
          Affymetrix Rat Gene 1.0 ST Array 10895873 - -
          Affymetrix Rat Gene 1.0 ST Array 10895879 - -
          Affymetrix Rat Gene 1.0 ST Array 10895880 - -
          Affymetrix Rat Gene 1.1 ST Array 10895866 - -
          Rat Genome 230 2.0 Array 1382026_PM_at - -
          Rat Genome 230 2.0 Array 1382026_at - -
          Affymetrix HT Rat Focus Array 1382026_PM_at - -
          Affymetrix HT Rat Focus Array 1382026_at - -
          Affymetrix Rat Clariom S Assay PSR0700006763.rn.1 TC0700000896.rn.1 RTA TC
          Rat Expression Set 230 1382026_at - -
          Rat Expression Set 230 1382026_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895866 - -
          Rat Genome U34 Set rc_AI237283_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20874280 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376877 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348820 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline