Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03061959_m1
          See other GNL3L GE Assays ›
          Gene Symbol:
          Gnl3l
          Gene Name:
          guanine nucleotide binding protein-like 3 (nucleolar)-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 20037558 - 20070505 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03061959_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnl3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081958.1 NP_001075427.1 9 - 10 824 60
          GenBank mRNA CA512184.1 - 4 - 5 448 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001556.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          ribosome biogenesis
          GTPase activity
          GTP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P175786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175786 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175786 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P175786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28968 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49996 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937545 - -
          Affymetrix Rat Gene 1.0 ST Array 10937550 - -
          Affymetrix Rat Gene 1.0 ST Array 10937542 - -
          Affymetrix Rat Gene 1.0 ST Array 10937551 - -
          Affymetrix Rat Gene 1.0 ST Array 10937546 - -
          Affymetrix Rat Gene 1.0 ST Array 10937553 - -
          Affymetrix Rat Gene 1.0 ST Array 10937543 - -
          Affymetrix Rat Gene 1.0 ST Array 10937547 - -
          Affymetrix Rat Gene 1.0 ST Array 10937549 - -
          Affymetrix Rat Gene 1.0 ST Array 10937548 - -
          Affymetrix Rat Gene 1.0 ST Array 10937541 - -
          Affymetrix Rat Gene 1.0 ST Array 10937544 - -
          Affymetrix Rat Gene 1.0 ST Array 10937552 - -
          Affymetrix Rat Gene 1.1 ST Array 10937540 - -
          Rat Genome 230 2.0 Array 1399067_PM_at - -
          Rat Genome 230 2.0 Array 1399067_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010059.rn.1 TC0X00001556.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010058.rn.1 TC0X00001556.rn.1 RTA TC
          Rat Expression Set 230 1399067_at - -
          Rat Expression Set 230 1399067_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937540 - -
          Rat Genome U34 Set rc_AI229702_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline