Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03302271_gH
          See other RPLP0 GE Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          Arbp
          Chromosome Location:
          Chr.12: 46791528 - 46794797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03302271_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022402.2 NP_071797.1 3 - 4 387 59
          GenBank mRNA BC062028.1 - 2 - 3 356 5623881 59
          DN931898.1 - 2 - 3 405 59
          FQ209537.1 - 3 - 4 387 59
          FQ209603.1 - 3 - 4 387 59
          FQ209620.1 - 2 - 3 372 59
          FQ209649.1 - 3 - 4 387 59
          FQ209757.1 - 3 - 4 388 59
          FQ210412.1 - 3 - 4 387 59
          FQ211152.1 - 3 - 4 387 59
          FQ211263.1 - 3 - 4 386 59
          FQ212554.1 - 3 - 4 387 59
          FQ212743.1 - 3 - 4 387 59
          FQ212875.1 - 3 - 4 387 59
          FQ212927.1 - 3 - 4 388 59
          FQ213085.1 - 3 - 4 387 59
          FQ213427.1 - 3 - 4 387 59
          FQ213510.1 - 3 - 4 387 59
          FQ213712.1 - 3 - 4 387 59
          FQ213847.1 - 3 - 4 388 59
          FQ214087.1 - 3 - 4 387 59
          FQ214354.1 - 3 - 4 387 59
          FQ214515.1 - 3 - 4 387 59
          FQ214553.1 - 3 - 4 388 59
          FQ214850.1 - 3 - 4 387 59
          FQ214997.1 - 3 - 4 387 59
          FQ215007.1 - 3 - 4 387 59
          FQ215548.1 - 3 - 4 389 59
          FQ215662.1 - 3 - 4 387 59
          FQ215682.1 - 3 - 4 387 59
          FQ215870.1 - 3 - 4 387 59
          FQ216120.1 - 3 - 4 388 59
          FQ216296.1 - 3 - 4 387 59
          FQ216389.1 - 3 - 4 388 59
          FQ216406.1 - 3 - 4 390 59
          FQ216618.1 - 3 - 4 387 59
          FQ216655.1 - 3 - 4 388 59
          FQ216733.1 - 3 - 4 389 59
          FQ216774.1 - 3 - 4 387 59
          FQ216813.1 - 3 - 4 386 59
          FQ216851.1 - 3 - 4 387 59
          FQ216913.1 - 3 - 4 388 59
          FQ217406.1 - 3 - 4 387 59
          FQ218561.1 - 3 - 4 387 59
          FQ218832.1 - 3 - 4 387 59
          FQ218943.1 - 3 - 4 387 59
          FQ219025.1 - 3 - 4 390 59
          FQ219184.1 - 3 - 4 391 59
          FQ219389.1 - 3 - 4 387 59
          FQ219457.1 - 3 - 4 387 59
          FQ219678.1 - 3 - 4 387 59
          FQ219815.1 - 3 - 4 387 59
          FQ219817.1 - 3 - 4 387 59
          FQ219837.1 - 3 - 4 387 59
          FQ219855.1 - 3 - 4 387 59
          FQ219923.1 - 3 - 4 387 59
          FQ220051.1 - 3 - 4 387 59
          FQ220077.1 - 3 - 4 387 59
          FQ220115.1 - 3 - 4 387 59
          FQ220181.1 - 3 - 4 387 59
          FQ220201.1 - 3 - 4 385 59
          FQ220213.1 - 3 - 4 387 59
          FQ220247.1 - 3 - 4 388 59
          FQ220284.1 - 3 - 4 387 59
          FQ220302.1 - 3 - 4 387 59
          FQ220338.1 - 3 - 4 387 59
          FQ220345.1 - 3 - 4 387 59
          FQ220370.1 - 3 - 4 387 59
          FQ220406.1 - 3 - 4 387 59
          FQ220413.1 - 3 - 4 387 59
          FQ220420.1 - 3 - 4 388 59
          FQ220433.1 - 3 - 4 384 59
          FQ220444.1 - 3 - 4 387 59
          FQ220445.1 - 3 - 4 383 59
          FQ220449.1 - 3 - 4 59
          FQ220500.1 - 3 - 4 387 59
          FQ220514.1 - 3 - 4 387 59
          FQ220533.1 - 3 - 4 419 59
          FQ220542.1 - 3 - 4 387 59
          FQ220545.1 - 3 - 4 387 59
          FQ220634.1 - 3 - 4 59
          FQ220654.1 - 3 - 4 387 59
          FQ220684.1 - 3 - 4 388 59
          FQ220690.1 - 3 - 4 389 59
          FQ220769.1 - 3 - 4 387 59
          FQ220801.1 - 3 - 4 387 59
          FQ220833.1 - 3 - 4 387 59
          FQ220838.1 - 3 - 4 387 59
          FQ220888.1 - 3 - 4 387 59
          FQ220932.1 - 3 - 4 388 59
          FQ221146.1 - 3 - 4 387 59
          FQ221380.1 - 3 - 4 387 59
          FQ221533.1 - 3 - 4 59
          FQ221556.1 - 3 - 4 385 59
          FQ222171.1 - 3 - 4 387 59
          FQ222181.1 - 3 - 4 387 59
          FQ222327.1 - 3 - 4 387 59
          FQ222772.1 - 3 - 4 387 59
          FQ222906.1 - 3 - 4 387 59
          FQ223121.1 - 3 - 4 387 59
          FQ223487.1 - 3 - 4 387 59
          FQ224547.1 - 3 - 4 387 59
          FQ224834.1 - 3 - 4 387 59
          FQ224846.1 - 3 - 4 387 59
          FQ225049.1 - 3 - 4 387 59
          FQ225170.1 - 3 - 4 387 59
          FQ225608.1 - 3 - 4 387 59
          FQ225654.1 - 3 - 4 389 59
          FQ225860.1 - 3 - 4 389 59
          FQ226306.1 - 3 - 4 389 59
          FQ227381.1 - 3 - 4 387 59
          FQ227522.1 - 3 - 4 382 59
          FQ227719.1 - 3 - 4 387 59
          FQ227837.1 - 3 - 4 387 59
          FQ227937.1 - 3 - 4 387 59
          FQ227941.1 - 3 - 4 387 59
          FQ227942.1 - 3 - 4 507 59
          FQ228548.1 - 3 - 4 387 59
          FQ228995.1 - 3 - 4 387 59
          FQ228996.1 - 3 - 4 387 59
          FQ229139.1 - 3 - 4 389 59
          FQ229159.1 - 3 - 4 387 59
          FQ229164.1 - 3 - 4 388 59
          FQ229208.1 - 3 - 4 387 59
          FQ229241.1 - 3 - 4 387 59
          FQ229244.1 - 3 - 4 387 59
          FQ229248.1 - 1 - 2 267 59
          FQ229272.1 - 3 - 4 387 59
          FQ229273.1 - 3 - 4 388 59
          FQ229303.1 - 3 - 4 387 59
          FQ229337.1 - 3 - 4 387 59
          FQ229352.1 - 3 - 4 387 59
          FQ229397.1 - 3 - 4 387 59
          FQ229408.1 - 3 - 4 388 59
          FQ229433.1 - 3 - 4 387 59
          FQ229448.1 - 3 - 4 387 59
          FQ229521.1 - 3 - 4 387 59
          FQ229547.1 - 3 - 4 387 59
          FQ229554.1 - 3 - 4 387 59
          FQ229566.1 - 3 - 4 383 59
          FQ229610.1 - 3 - 4 387 59
          FQ229626.1 - 3 - 4 384 59
          FQ229726.1 - 3 - 4 387 59
          FQ229740.1 - 3 - 4 387 59
          FQ229742.1 - 3 - 4 387 59
          FQ229776.1 - 3 - 4 387 59
          FQ229808.1 - 3 - 4 387 59
          FQ229815.1 - 3 - 4 387 59
          FQ229822.1 - 3 - 4 387 59
          FQ229830.1 - 3 - 4 389 59
          FQ229883.1 - 3 - 4 389 59
          FQ229901.1 - 3 - 4 387 59
          FQ229932.1 - 1 - 2 285 59
          FQ229952.1 - 3 - 4 387 59
          FQ229974.1 - 3 - 4 387 59
          FQ230055.1 - 3 - 4 386 59
          FQ230127.1 - 3 - 4 388 59
          FQ230943.1 - 3 - 4 387 59
          FQ231445.1 - 3 - 4 389 59
          FQ231525.1 - 3 - 4 387 59
          FQ231591.1 - 3 - 4 389 59
          FQ231710.1 - 3 - 4 388 59
          FQ231733.1 - 3 - 4 388 59
          FQ231967.1 - 3 - 4 387 59
          FQ232029.1 - 3 - 4 388 59
          FQ232154.1 - 3 - 4 387 59
          FQ232165.1 - 3 - 4 387 59
          FQ232310.1 - 3 - 4 386 59
          FQ232319.1 - 3 - 4 388 59
          FQ232951.1 - 3 - 4 387 59
          FQ233152.1 - 3 - 4 386 59
          FQ233377.1 - 3 - 4 387 59
          FQ233473.1 - 3 - 4 387 59
          FQ233732.1 - 3 - 4 387 59
          FQ233737.1 - 3 - 4 387 59
          FQ233808.1 - 3 - 4 387 59
          FQ234195.1 - 3 - 4 387 59
          FQ234613.1 - 3 - 4 387 59
          FQ234733.1 - 3 - 4 387 59
          FQ234863.1 - 3 - 4 387 59
          FQ235063.1 - 3 - 4 387 59
          FQ235079.1 - 3 - 4 387 59
          FQ235122.1 - 3 - 4 387 59
          FQ235259.1 - 3 - 4 387 59
          X15096.1 - 3 - 4 387 59
          Z29530.1 - 2 - 3 343 59

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp0

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          ribosome biogenesis
          cellular response to interleukin-4
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1200014849.rn.1 TC1200001737.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline