Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03302274_m1
          See other PPIB GE Assays ›
          Gene Symbol:
          Ppib
          Gene Name:
          peptidylprolyl isomerase B
          Gene Aliases:
          CypB, Scylp
          Chromosome Location:
          Chr.8: 71719681 - 71725538 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03302274_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppib
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022536.2 NP_071981.1 4 - 5 527 70
          GenBank mRNA AF071225.1 - 4 - 5 527 70
          BC061971.1 - 4 - 5 556 5623342 70
          FQ211358.1 - 4 - 5 531 70
          FQ212031.1 - 4 - 5 556 70
          FQ219792.1 - 4 - 5 565 70
          FQ219926.1 - 4 - 5 577 70
          FQ221020.1 - 4 - 5 580 70
          FQ221248.1 - 4 - 5 575 70
          FQ221362.1 - 4 - 5 554 70
          FQ222696.1 - 4 - 5 689 70
          FQ223162.1 - 4 - 5 574 70
          FQ228087.1 - 4 - 5 570 70
          FQ229174.1 - 4 - 5 577 70
          FQ229214.1 - 4 - 5 571 70
          FQ231656.1 - 4 - 5 570 70
          FQ232077.1 - 4 - 5 566 70
          FQ232920.1 - 4 - 5 580 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppib

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          positive regulation of multicellular organism growth
          positive regulation by host of viral process
          positive regulation by host of viral genome replication
          protein stabilization
          nuclear transport
          bone development
          chaperone-mediated protein folding
          regulation of post-translational protein modification
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          protein complex binding
          peptide binding
          poly(A) RNA binding
          RNA polymerase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13976 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13976 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13976 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700148611 - -
          Agilent Rat cDNA Microarray (G4105A) 700148611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911040 - -
          Affymetrix Rat Gene 1.0 ST Array 10911039 - -
          Affymetrix Rat Gene 1.0 ST Array 10911038 - -
          Affymetrix Rat Gene 1.0 ST Array 10911036 - -
          Affymetrix Rat Gene 1.0 ST Array 10911037 - -
          Affymetrix Rat Gene 1.0 ST Array 10911041 - -
          Affymetrix Rat Gene 1.1 ST Array 10911035 - -
          Rat Genome 230 2.0 Array 1369934_at - -
          Rat Genome 230 2.0 Array 1397502_at - -
          Rat Genome 230 2.0 Array 1369934_PM_at - -
          Rat Genome 230 2.0 Array 1397502_PM_at - -
          Affymetrix HT Rat Focus Array 1369934_at - -
          Affymetrix HT Rat Focus Array 1369934_PM_at - -
          Rat Expression Set 230 1369934_at - -
          Rat Expression Set 230 1369934_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911035 - -
          Rat Genome U34 Set AF071225_at - -
          Rat Genome U34 Set rc_AA818487_s_at - -
          Rat Genome U34 Set rc_AA818487_s_at - -
          Rat Genome U34 Set AF071225_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22163589 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370397 - -
          RatRef-12 Expression BeadChip ILMN_63100 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline