Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03399817_g1
          See other TRNT1 GE Assays ›
          Gene Symbol:
          Trnt1
          Gene Name:
          tRNA nucleotidyl transferase, CCA-adding, 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 138855514 - 138869217 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03399817_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trnt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024261.2 NP_001019432.1 8 1198 71
          XM_008763191.1 XP_008761413.1 8 1247 71
          XM_008763192.1 XP_008761414.1 8 1203 71
          XM_008763193.1 XP_008761415.1 8 1157 71
          GenBank mRNA BC095841.1 - 7 1123 7387807 71
          BC105612.1 - 8 1198 7135310 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trnt1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA 3'-end processing
          tRNA binding
          tRNA adenylyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143856 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143856 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P521545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17527 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38313 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6638 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725154 - -
          Affymetrix Rat Gene 1.0 ST Array 10857539 - -
          Affymetrix Rat Gene 1.0 ST Array 10857540 - -
          Affymetrix Rat Gene 1.0 ST Array 10725153 - -
          Affymetrix Rat Gene 1.0 ST Array 10725152 - -
          Affymetrix Rat Gene 1.0 ST Array 10725151 - -
          Affymetrix Rat Gene 1.1 ST Array 10725150 - -
          Affymetrix Rat Gene 1.1 ST Array 10857538 - -
          Rat Genome 230 2.0 Array 1383144_at - -
          Rat Genome 230 2.0 Array 1383144_PM_at - -
          Affymetrix HT Rat Focus Array 1383144_PM_at - -
          Affymetrix HT Rat Focus Array 1383144_at - -
          Affymetrix Rat Clariom S Assay PSR0400010643.rn.1 TC0400001524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400010642.rn.1 TC0400001524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400010644.rn.1 TC0400001524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400010638.rn.1 TC0400001524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400010641.rn.1 TC0400001524.rn.1 RTA TC
          Rat Expression Set 230 1383144_PM_at - -
          Rat Expression Set 230 1383144_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857538 - -
          Affymetrix Rat Gene 1.0 ST Array 10725150 - -
          Rat Genome U34 Set rc_AI012382_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22219441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline