Search
Search

LOC690784| Gene Symbol: LOC690784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044299.1 | NP_001037764.1 | 2 - 3 | 790 | 68 | |
| XM_008763427.1 | XP_008761649.1 | 1 - 2 | 1896 | 68 | ||
| GenBank mRNA | BC082012.1 | - | 2 - 3 | 790 | 7132059 | 68 |
Set Membership: |
|