Search
Search

LOC685406| Gene Symbol: LOC685406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047977.1 | NP_001041442.1 | 2 - 3 | 104 | 69 | |
| GenBank mRNA | AY387048.1 | - | 2 - 3 | 104 | 69 | |
Set Membership: |
|