Search
Search
| Gene Symbol: Jrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104612.2 | NP_001098082.1 | 2 | 2005 | 76 | |
| XM_006241728.2 | XP_006241790.1 | 2 | 2587 | 76 | ||
| XM_008765569.1 | XP_008763791.1 | 2 | 2149 | 76 | ||
| GenBank mRNA | BC152551.1 | - | 1 | 1749 | 7133465 | 76 |
Set Membership: |
|