Search
Search

Olr1469
LOC100909534
Olr1482| Gene Symbol: Olr1469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212527.1 | NP_997692.1 | 1 | 149 | 137 | |
| GenBank mRNA | Y07557.1 | - | 1 | 149 | 137 | |
| Gene Symbol: LOC100909534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750852.1 | XP_003750900.1 | 1 | 149 | 137 | |
| Gene Symbol: Olr1482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768133.1 | XP_008766355.1 | 1 | 149 | 137 | |
Set Membership: |
|