Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03467157_gH
          See other RPLP1 GE Assays ›
          Gene Symbol:
          Rplp1
          Gene Name:
          ribosomal protein, large, P1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03467157_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007604.2 NP_001007605.1 2 - 3 264 96
          GenBank mRNA BC058151.1 - 2 - 3 267 6888011 96
          FQ209966.1 - 2 - 3 266 96
          FQ210237.1 - 2 - 3 263 96
          FQ210864.1 - 2 - 3 263 96
          FQ210988.1 - 2 - 3 266 96
          FQ211097.1 - 2 - 3 266 96
          FQ211327.1 - 2 - 3 264 96
          FQ211648.1 - 2 - 3 263 96
          FQ212351.1 - 2 - 3 263 96
          FQ214973.1 - 2 - 3 266 96
          FQ215065.1 - 2 - 3 263 96
          FQ216905.1 - 2 - 3 267 96
          FQ216921.1 - 2 - 3 266 96
          FQ216959.1 - 2 - 3 262 96
          FQ216979.1 - 2 - 3 266 96
          FQ217313.1 - 2 - 3 264 96
          FQ220167.1 - 2 - 3 264 96
          FQ220212.1 - 2 - 3 265 96
          FQ221027.1 - 2 - 3 264 96
          FQ221120.1 - 2 - 3 264 96
          FQ221137.1 - 2 - 3 263 96
          FQ221201.1 - 2 - 3 266 96
          FQ221359.1 - 2 - 3 264 96
          FQ221455.1 - 2 - 3 268 96
          FQ221534.1 - 2 - 3 261 96
          FQ221567.1 - 2 - 3 266 96
          FQ221595.1 - 2 - 3 264 96
          FQ221620.1 - 2 - 3 263 96
          FQ221717.1 - 2 - 3 263 96
          FQ221756.1 - 2 - 3 263 96
          FQ221784.1 - 2 - 3 263 96
          FQ221894.1 - 2 - 3 263 96
          FQ221969.1 - 2 - 3 263 96
          FQ221980.1 - 2 - 3 263 96
          FQ222031.1 - 2 - 3 263 96
          FQ222178.1 - 2 - 3 263 96
          FQ222307.1 - 2 - 3 265 96
          FQ222468.1 - 2 - 3 265 96
          FQ222559.1 - 2 - 3 263 96
          FQ222608.1 - 2 - 3 266 96
          FQ222801.1 - 2 - 3 264 96
          FQ222805.1 - 2 - 3 263 96
          FQ222816.1 - 2 - 3 264 96
          FQ222879.1 - 2 - 3 259 96
          FQ222923.1 - 2 - 3 264 96
          FQ222965.1 - 2 - 3 263 96
          FQ222966.1 - 2 - 3 262 96
          FQ223013.1 - 2 - 3 264 96
          FQ223016.1 - 2 - 3 264 96
          FQ223066.1 - 2 - 3 265 96
          FQ223344.1 - 2 - 3 264 96
          FQ223388.1 - 2 - 3 263 96
          FQ223470.1 - 2 - 3 266 96
          FQ223512.1 - 2 - 3 264 96
          FQ223537.1 - 2 - 3 264 96
          FQ223567.1 - 2 - 3 263 96
          FQ223591.1 - 2 - 3 263 96
          FQ223674.1 - 2 - 3 245 96
          FQ223769.1 - 2 - 3 264 96
          FQ224482.1 - 2 - 3 233 96
          FQ224625.1 - 2 - 3 266 96
          FQ224766.1 - 2 - 3 264 96
          FQ228323.1 - 2 - 3 263 96
          FQ228361.1 - 2 - 3 263 96
          FQ228390.1 - 2 - 3 264 96
          FQ228490.1 - 2 - 3 264 96
          FQ228523.1 - 2 - 3 263 96
          FQ228528.1 - 2 - 3 260 96
          FQ228539.1 - 2 - 3 263 96
          FQ228605.1 - 2 - 3 267 96
          FQ228792.1 - 2 - 3 266 96
          FQ228990.1 - 2 - 3 270 96
          FQ229009.1 - 2 - 3 263 96
          FQ229050.1 - 2 - 3 263 96
          FQ229318.1 - 2 - 3 264 96
          FQ230089.1 - 2 - 3 264 96
          X15097.1 - 2 - 3 234 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp1

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749143 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027003 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027003 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027003 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P621736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918028 - -
          Affymetrix Rat Gene 1.0 ST Array 10918029 - -
          Affymetrix Rat Gene 1.0 ST Array 10797171 - -
          Affymetrix Rat Gene 1.0 ST Array 10797172 - -
          Affymetrix Rat Gene 1.0 ST Array 10797170 - -
          Affymetrix Rat Gene 1.0 ST Array 10918030 - -
          Affymetrix Rat Gene 1.1 ST Array 10918027 - -
          Affymetrix Rat Gene 1.1 ST Array 10797169 - -
          Rat Genome 230 2.0 Array 1371307_PM_at - -
          Rat Genome 230 2.0 Array 1371307_at - -
          Affymetrix HT Rat Focus Array 1371307_PM_at - -
          Affymetrix HT Rat Focus Array 1371307_at - -
          Affymetrix Rat Clariom S Assay PSR0800021967.rn.1 TC0800002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021964.rn.1 TC0800002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021963.rn.1 TC0800002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800021966.rn.1 TC0800002707.rn.1 RTA TC
          Rat Expression Set 230 1371307_at - -
          Rat Expression Set 230 1371307_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918027 - -
          Affymetrix Rat Gene 1.0 ST Array 10797169 - -
          Rat Genome U34 Set rc_AA945636_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21663420 - -
          ABI Rat Genome Survey Microarray 21845694 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360590 - -
          RatRef-12 Expression BeadChip ILMN_51310 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline