Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03648292_m1
          See other RGD1564114 GE Assays ›
          Gene Symbol:
          RGD1564114
          Gene Name:
          similar to FLJ46082 protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 7508500 - 7632311 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03648292_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1564114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127563.1 NP_001121035.1 5 - 6 759 61
          GenBank mRNA BC166521.1 - 5 - 6 759 7133997 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002790.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          structural protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141727 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102012 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089265 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96723 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844076 - -
          Affymetrix Rat Gene 1.0 ST Array 10844079 - -
          Affymetrix Rat Gene 1.0 ST Array 10844077 - -
          Affymetrix Rat Gene 1.0 ST Array 10844078 - -
          Affymetrix Rat Gene 1.0 ST Array 10844075 - -
          Affymetrix Rat Gene 1.0 ST Array 10844081 - -
          Affymetrix Rat Gene 1.1 ST Array 10844074 - -
          Rat Genome 230 2.0 Array 1375992_PM_at - -
          Rat Genome 230 2.0 Array 1375992_at - -
          Affymetrix HT Rat Focus Array 1375992_PM_at - -
          Affymetrix HT Rat Focus Array 1375992_at - -
          Affymetrix Rat Clariom S Assay PSR0300021235.rn.1 TC0300002790.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300021236.rn.1 TC0300002790.rn.1 RTA TC
          Rat Expression Set 230 1375992_at - -
          Rat Expression Set 230 1375992_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844074 - -
          Rat Genome U34 Set rc_AI137751_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0