Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03810033_s1
          See other ZFP329 GE Assays ›
          Gene Symbol:
          Zfp329
          Gene Name:
          zinc finger protein 329
          Gene Aliases:
          RGD1565029
          Chromosome Location:
          Chr.1: 65781506 - 65800492 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03810033_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp329
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107477.2 NP_001100947.1 2 1549 111
          XM_006228109.2 XP_006228171.1 2 1842 111
          XM_006228110.2 XP_006228172.1 1 1534 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P212922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212922 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212922 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P212922 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719087 - -
          Affymetrix Rat Gene 1.1 ST Array 10719086 - -
          Rat Genome 230 2.0 Array 1384118_at - -
          Rat Genome 230 2.0 Array 1397467_at - -
          Rat Genome 230 2.0 Array 1394969_at - -
          Rat Genome 230 2.0 Array 1397467_PM_at - -
          Rat Genome 230 2.0 Array 1394969_PM_at - -
          Rat Genome 230 2.0 Array 1384118_PM_at - -
          Affymetrix HT Rat Focus Array 1384118_PM_at - -
          Affymetrix HT Rat Focus Array 1384118_at - -
          Affymetrix Rat Clariom S Assay PSR0100005000.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005001.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004999.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004995.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004997.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004998.rn.1 TC0100000746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100004996.rn.1 TC0100000746.rn.1 RTA TC
          Rat Expression Set 230 1394969_at - -
          Rat Expression Set 230 1394969_PM_at - -
          Rat Expression Set 230 1397467_PM_at - -
          Rat Expression Set 230 1384118_at - -
          Rat Expression Set 230 1384118_PM_at - -
          Rat Expression Set 230 1397467_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719086 - -
          Rat Genome U34 Set rc_AA956308_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22070028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline