Search
Search

Trim43a| Gene Symbol: Trim43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142944.1 | NP_001136416.1 | 3 - 4 | 739 | 78 | |
| XM_006243512.2 | XP_006243574.1 | 4 - 5 | 979 | 78 | ||
Set Membership: |
|