Search
Search
| Gene Symbol: Tlr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144779.2 | NP_001138251.2 | 2 | 2837 | 78 | |
| GenBank mRNA | FJ539013.1 | - | 1 | 2748 | 78 | |
| FQ230537.1 | - | 2 | 2837 | 78 | ||
Set Membership: |
|