Search
Search
| Gene Symbol: Tlr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146035.1 | NP_001139507.1 | 1 | 2350 | 77 | |
| XM_006251002.2 | XP_006251064.1 | 2 | 2456 | 77 | ||
| GenBank mRNA | FJ755908.1 | - | 1 | 2350 | 77 | |
Set Membership: |
|