Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03928990_g1
          See other B2M GE Assays ›
          Gene Symbol:
          B2m Hbb LOC689064 Pfdn1 Rn18s Rn45s Rpl4
          Gene Name:
          beta-2 microglobulin
          hemoglobin, beta
          beta-globin
          prefoldin subunit 1
          18S ribosomal RNA
          45S pre-ribosomal RNA
          ribosomal protein L4
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168971269 - 168972680 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168964202 - 168965568 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 29233987 - 29290480 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 46643222 - 46645093 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 46643222 - 46655563 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 69121682 - 69126805 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          Assay ID Rn03928990_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ219455.1 - 152 61
          Gene Symbol: Hbb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ235254.1 - 152 61
          Gene Symbol: LOC689064
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ228130.1 - 152 61
          Gene Symbol: Pfdn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ227006.1 - 152 61
          FQ234841.1 - 152 61
          Gene Symbol: Rn18s
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046237.1 1 152 61
          Gene Symbol: Rn45s
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046239.1 1 152 61
          Gene Symbol: Rpl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FQ212694.1 - 152 61

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548937 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156319 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156319 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724312 - -
          Affymetrix Rat Gene 1.0 ST Array 10724314 - -
          Affymetrix Rat Gene 1.0 ST Array 10724313 - -
          Affymetrix Rat Gene 1.1 ST Array 10724311 - -
          Rat Genome 230 2.0 Array 1371102_x_at - -
          Rat Genome 230 2.0 Array 1371245_a_at - -
          Rat Genome 230 2.0 Array 1371102_PM_x_at - -
          Rat Genome 230 2.0 Array 1371245_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371245_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371245_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400004258.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004255.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004261.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004262.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004253.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004257.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004263.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004250.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004260.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004256.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004252.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004259.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004254.rn.1 TC1400000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400004251.rn.1 TC1400000572.rn.1 RTA TC
          Rat Expression Set 230 1371102_PM_x_at - -
          Rat Expression Set 230 1371102_x_at - -
          Rat Expression Set 230 1371245_a_at - -
          Rat Expression Set 230 1371245_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724311 - -
          Rat Genome U34 Set rc_AI179576_s_at - -
          Rat Genome U34 Set rc_AI011943_at - -
          Rat Genome U34 Set rc_AI012182_s_at - -
          Rat Genome U34 Set rc_AI177992_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline