Search
Search

RGD1561230| Gene Symbol: RGD1561230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106968.1 | NP_001100438.1 | 1 | 350 | 179 | |
| XM_006257206.2 | XP_006257268.1 | 2 | 477 | 179 | ||
| XM_008773374.1 | XP_008771596.1 | 2 | 477 | 179 | ||
Set Membership: |
|