Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04219426_mH
          See other GDI2 GE Assays ›
          Gene Symbol:
          Gdi2
          Gene Name:
          GDP dissociation inhibitor 2
          Gene Aliases:
          Gdi3
          Chromosome Location:
          Chr.17: 70299177 - 70325864 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04219426_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdi2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017276.2 NP_058972.2 8 - 9 1087 71
          GenBank mRNA BC061767.1 - 8 - 9 1087 5621162 71
          X74401.1 - 8 - 9 1017 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002070.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gdi2

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          Rab GDP-dissociation inhibitor activity
          small GTPase binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P390454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P390454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P390454 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P390454 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P390454 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484826 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700530402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92851 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39127 - -
          Agilent Rat cDNA Microarray (G4105A) 700530402 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799385 - -
          Affymetrix Rat Gene 1.0 ST Array 10799384 - -
          Affymetrix Rat Gene 1.0 ST Array 10799395 - -
          Affymetrix Rat Gene 1.0 ST Array 10799390 - -
          Affymetrix Rat Gene 1.0 ST Array 10799394 - -
          Affymetrix Rat Gene 1.0 ST Array 10799396 - -
          Affymetrix Rat Gene 1.0 ST Array 10799389 - -
          Affymetrix Rat Gene 1.0 ST Array 10799387 - -
          Affymetrix Rat Gene 1.0 ST Array 10799391 - -
          Affymetrix Rat Gene 1.0 ST Array 10799388 - -
          Affymetrix Rat Gene 1.0 ST Array 10799393 - -
          Affymetrix Rat Gene 1.0 ST Array 10799392 - -
          Affymetrix Rat Gene 1.0 ST Array 10799386 - -
          Affymetrix Rat Gene 1.1 ST Array 10799383 - -
          Rat Genome 230 2.0 Array 1367460_PM_at - -
          Rat Genome 230 2.0 Array 1367460_at - -
          Rat Genome 230 2.0 Array 1398822_PM_at - -
          Rat Genome 230 2.0 Array 1398822_at - -
          Affymetrix HT Rat Focus Array 1367460_PM_at - -
          Affymetrix HT Rat Focus Array 1367460_at - -
          Affymetrix HT Rat Focus Array 1398822_at - -
          Affymetrix HT Rat Focus Array 1398822_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700014236.rn.1 TC1700002070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014239.rn.1 TC1700002070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014238.rn.1 TC1700002070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700014237.rn.1 TC1700002070.rn.1 RTA TC
          Rat Expression Set 230 1367460_PM_at - -
          Rat Expression Set 230 1398822_PM_at - -
          Rat Expression Set 230 1367460_at - -
          Rat Expression Set 230 1398822_at - -
          Rat Expression Set 230 1367460_at - -
          Rat Expression Set 230 1367460_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10799383 - -
          Rat Genome U34 Set X74401_at - -
          Rat Genome U34 Set rc_AI105149_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21686201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361811 - -
          RatRef-12 Expression BeadChip ILMN_51492 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline