Search
Search

Hist1h4b| Gene Symbol: Hist1h4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022686.2 | NP_073177.1 | 1 - 2 | 326 | 98 | |
| GenBank mRNA | AY936209.1 | - | 1 - 2 | 326 | 98 | |
| BC100619.1 | - | 1 - 2 | 330 | 7938313 | 98 | |
Set Membership: |
|