Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04219720_m1
          See other SPINK1 GE Assays ›
          Gene Symbol:
          Spink1
          Gene Name:
          serine peptidase inhibitor, Kazal type 1
          Gene Aliases:
          Psti, Spink3
          Chromosome Location:
          Chr.18: 38177441 - 38185812 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04219720_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spink1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152936.1 NP_690919.1 3 - 4 252 93
          GenBank mRNA FQ209906.1 - 3 - 4 233 93
          FQ210000.1 - 3 - 4 266 93
          FQ210029.1 - 3 - 4 265 93
          FQ210105.1 - 3 - 4 270 93
          FQ210138.1 - 3 - 4 265 93
          FQ210344.1 - 3 - 4 265 93
          FQ210442.1 - 3 - 4 265 93
          FQ210447.1 - 3 - 4 265 93
          FQ210555.1 - 3 - 4 230 93
          FQ210766.1 - 3 - 4 267 93
          FQ210836.1 - 3 - 4 267 93
          FQ210933.1 - 3 - 4 267 93
          FQ211214.1 - 3 - 4 269 93
          FQ211434.1 - 3 - 4 251 93
          FQ218107.1 - 3 - 4 267 93
          FQ218122.1 - 3 - 4 266 93
          FQ218165.1 - 3 - 4 267 93
          FQ218208.1 - 3 - 4 263 93
          FQ218422.1 - 3 - 4 265 93
          M27883.1 - 3 - 4 252 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001365.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spink1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of nitric oxide mediated signal transduction
          negative regulation of endopeptidase activity
          negative regulation of peptidyl-tyrosine phosphorylation
          regulation of acrosome reaction
          negative regulation of calcium ion import
          negative regulation of serine-type endopeptidase activity
          regulation of store-operated calcium entry
          serine-type endopeptidase inhibitor activity
          calmodulin binding
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P313452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50411 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313452 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313452 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P313452 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P313452 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701191029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28254 - -
          Agilent Rat cDNA Microarray (G4105A) 701191029 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10804291 - -
          Affymetrix Rat Gene 1.0 ST Array 10804290 - -
          Affymetrix Rat Gene 1.0 ST Array 10804289 - -
          Affymetrix Rat Gene 1.1 ST Array 10804288 - -
          Rat Genome 230 2.0 Array 1387967_at - -
          Rat Genome 230 2.0 Array 1387967_PM_at - -
          Affymetrix HT Rat Focus Array 1387967_at - -
          Affymetrix HT Rat Focus Array 1387967_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800009848.rn.1 TC1800001365.rn.1 RTA TC
          Rat Expression Set 230 1387967_PM_at - -
          Rat Expression Set 230 1387967_at - -
          Affymetrix Rat Gene 1.0 ST Array 10804288 - -
          Rat Genome U34 Set rc_AA858673_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21594725 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371222 - -
          RatRef-12 Expression BeadChip ILMN_59263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline