Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04222618_gH
          See other LOC500959 GE Assays ›
          Gene Symbol:
          LOC500959 LOC100911515 Tpi1
          Gene Name:
          triosephosphate isomerase
          triosephosphate isomerase-like
          triosephosphate isomerase 1
          Gene Aliases:
          -
          -
          Tpi
          Chromosome Location:
          Chr.8: 26341789 - 26342573 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 20213596 - 20217278 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 157328375 - 157331905 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn04222618_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC500959
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033072.1 NP_001028244.1 1 48 100
          GenBank mRNA AY461585.1 - 1 48 100
          Gene Symbol: LOC100911515
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750654.3 XP_003750702.2 1 207 100
          Gene Symbol: Tpi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022922.2 NP_075211.2 1 51 100
          GenBank mRNA BC126087.1 - 1 51 8368624 100
          FQ214607.1 - 1 136 100
          FQ214670.1 - 1 59 100
          FQ214880.1 - 1 64 100
          FQ215529.1 - 1 63 100
          FQ215810.1 - 1 64 100
          FQ216584.1 - 1 63 100
          FQ224880.1 - 1 62 100
          FQ224920.1 - 1 66 100
          FQ224970.1 - 1 59 100
          L36250.1 - 1 51 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the AY461585.1,BC126087.1,L36250.1,NM_001033072.1,NM_022922.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC500959
          Click here to find siRNAs for Gene LOC100911515
          Click here to find siRNAs for Gene Tpi1

          Panther Classification:

          Molecular Function -

          isomerase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          multicellular organism development
          glyceraldehyde-3-phosphate metabolic process
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          pentose-phosphate shunt
          triose-phosphate isomerase activity
          isomerase activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011704 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P111225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011704 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011704 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P111225 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915838 - -
          Affymetrix Rat Gene 1.0 ST Array 10798022 - -
          Affymetrix Rat Gene 1.1 ST Array 10915837 - -
          Affymetrix Rat Gene 1.1 ST Array 10798021 - -
          Affymetrix Rat Clariom S Assay PSR0800017324.rn.1 TC0800002121.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10915837 - -
          Affymetrix Rat Gene 1.0 ST Array 10798021 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361472 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355142 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370343 - -
          RatRef-12 Expression BeadChip ILMN_54573 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0