Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04222789_gH
          See other RPL7 GE Assays ›
          Gene Symbol:
          Rpl7
          Gene Name:
          ribosomal protein L7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 2633517 - 2636525 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04222789_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100534.1 NP_001094004.1 3 - 4 575 78
          GenBank mRNA BC159423.1 - 3 - 4 463 78
          BC159431.1 - 3 - 4 575 9091211 78
          FQ211668.1 - 2 - 3 270 78
          FQ217699.1 - 2 - 3 270 78
          FQ219786.1 - 2 - 3 270 78
          FQ220709.1 - 2 - 3 270 78
          FQ220767.1 - 2 - 3 270 78
          FQ221206.1 - 2 - 3 269 78
          FQ221432.1 - 2 - 3 270 78
          FQ221889.1 - 2 - 3 270 78
          FQ222683.1 - 2 - 3 270 78
          FQ223086.1 - 2 - 3 270 78
          FQ223813.1 - 2 - 3 270 78
          FQ224905.1 - 2 - 3 270 78
          FQ225844.1 - 2 - 3 270 78
          FQ226679.1 - 2 - 3 270 78
          FQ228074.1 - 2 - 3 270 78
          FQ228176.1 - 2 - 3 270 78
          FQ229585.1 - 2 - 3 270 78
          FQ229680.1 - 2 - 3 270 78
          FQ229824.1 - 2 - 3 270 78
          FQ232074.1 - 2 - 3 270 78
          FQ232489.1 - 2 - 3 270 78
          FQ232741.1 - 2 - 3 270 78
          FQ234236.1 - 2 - 3 270 78
          FQ235314.1 - 2 - 3 270 78
          M17422.1 - 4 - 5 596 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          ribosomal large subunit biogenesis
          DNA binding
          RNA binding
          mRNA binding
          protein homodimerization activity
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0500000119.rn.1 TC0500000027.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000120.rn.1 TC0500000027.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000121.rn.1 TC0500000027.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000123.rn.1 TC0500000027.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline