Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04222958_g1
          See other HPCA GE Assays ›
          Gene Symbol:
          Hpca
          Gene Name:
          hippocalcin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 147295121 - 147305753 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04222958_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpca
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017122.1 NP_058818.1 2 - 3 660 88
          XM_006238920.1 XP_006238982.1 3 - 4 605 88
          XM_006238921.2 XP_006238983.1 3 - 4 562 88
          GenBank mRNA AY442172.1 - 2 - 3 487 88
          BC087632.1 - 2 - 3 515 7319524 88
          D12573.1 - 2 - 3 660 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003657.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpca

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          positive regulation of phospholipase activity
          response to organic cyclic compound
          calcium-mediated signaling
          negative regulation of guanylate cyclase activity
          positive regulation of adenylate cyclase activity
          inner ear development
          retina development in camera-type eye
          cellular response to electrical stimulus
          positive regulation of protein targeting to membrane
          response to ketamine
          response to L-glutamate
          actin binding
          calcium ion binding
          protein binding
          kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P165964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165964 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165964 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P165964 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6791 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879968 - -
          Affymetrix Rat Gene 1.0 ST Array 10879965 - -
          Affymetrix Rat Gene 1.0 ST Array 10879967 - -
          Affymetrix Rat Gene 1.0 ST Array 10879966 - -
          Affymetrix Rat Gene 1.0 ST Array 10879964 - -
          Affymetrix Rat Gene 1.1 ST Array 10879963 - -
          Rat Genome 230 2.0 Array 1387387_PM_at - -
          Rat Genome 230 2.0 Array 1387387_at - -
          Affymetrix HT Rat Focus Array 1387387_PM_at - -
          Affymetrix HT Rat Focus Array 1387387_at - -
          Affymetrix Rat Clariom S Assay PSR0500027238.rn.1 TC0500003657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500027239.rn.1 TC0500003657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500027240.rn.1 TC0500003657.rn.1 RTA TC
          Rat Expression Set 230 1387387_PM_at - -
          Rat Expression Set 230 1387387_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879963 - -
          Rat Genome U34 Set D12573_at - -
          Rat Genome U34 Set D12573_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21333675 - -
          ABI Rat Genome Survey Microarray 21646565 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376948 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline