Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223824_s1
          See other ARPC5L GE Assays ›
          Gene Symbol:
          Arpc5l
          Gene Name:
          actin related protein 2/3 complex, subunit 5-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 23301455 - 23309077 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04223824_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arpc5l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037767.2 NP_001032856.2 4 1141 93
          GenBank mRNA BC127445.1 - 4 1070 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arpc5l

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          Arp2/3 complex-mediated actin nucleation
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139113 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064523 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139113 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064521 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139113 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064521 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064523 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139113 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064521 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1058222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P345102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836013 - -
          Affymetrix Rat Gene 1.0 ST Array 10836012 - -
          Affymetrix Rat Gene 1.0 ST Array 10927679 - -
          Affymetrix Rat Gene 1.0 ST Array 10836014 - -
          Affymetrix Rat Gene 1.0 ST Array 10836011 - -
          Affymetrix Rat Gene 1.1 ST Array 10927678 - -
          Affymetrix Rat Gene 1.1 ST Array 10836010 - -
          Rat Genome 230 2.0 Array 1375641_PM_at - -
          Rat Genome 230 2.0 Array 1375641_at - -
          Rat Genome 230 2.0 Array 1382016_PM_at - -
          Rat Genome 230 2.0 Array 1382016_at - -
          Affymetrix HT Rat Focus Array 1382016_PM_at - -
          Affymetrix HT Rat Focus Array 1382016_at - -
          Affymetrix HT Rat Focus Array 1375641_at - -
          Affymetrix HT Rat Focus Array 1375641_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300004096.rn.1 TC0300000476.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300004095.rn.1 TC0300000476.rn.1 RTA TC
          Rat Expression Set 230 1382016_PM_at - -
          Rat Expression Set 230 1375641_PM_at - -
          Rat Expression Set 230 1375641_at - -
          Rat Expression Set 230 1382016_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836010 - -
          Rat Genome U34 Set rc_H32977_at - -
          Rat Genome U34 Set rc_AI007722_at - -
          Rat Genome U34 Set H32978_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21854419 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359645 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline