Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223839_u1
          See other ACAA1B GE Assays ›
          Gene Symbol:
          Acaa1b
          Gene Name:
          acetyl-Coenzyme A acyltransferase 1B
          Gene Aliases:
          RGD1562373
          Chromosome Location:
          Chr.8: 127836237 - 127845028 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04223839_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acaa1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040019.1 NP_001035108.1 12 1454 112
          GenBank mRNA J02749.1 - 12 1454 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          acetyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          fatty acid metabolic process
          fatty acid beta-oxidation
          metabolic process
          bile acid metabolic process
          acetyl-CoA C-acyltransferase activity
          protein binding
          palmitoyl-CoA oxidase activity
          transferase activity, transferring acyl groups other than amino-acyl groups

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029333 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029335 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029335 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029333 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029333 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029335 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161052 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920821 - -
          Affymetrix Rat Gene 1.0 ST Array 10920828 - -
          Affymetrix Rat Gene 1.0 ST Array 10920824 - -
          Affymetrix Rat Gene 1.0 ST Array 10920822 - -
          Affymetrix Rat Gene 1.0 ST Array 10914206 - -
          Affymetrix Rat Gene 1.0 ST Array 10920829 - -
          Affymetrix Rat Gene 1.0 ST Array 10920827 - -
          Affymetrix Rat Gene 1.0 ST Array 10920832 - -
          Affymetrix Rat Gene 1.0 ST Array 10920825 - -
          Affymetrix Rat Gene 1.0 ST Array 10920823 - -
          Affymetrix Rat Gene 1.0 ST Array 10920831 - -
          Affymetrix Rat Gene 1.0 ST Array 10920826 - -
          Affymetrix Rat Gene 1.0 ST Array 10914207 - -
          Affymetrix Rat Gene 1.0 ST Array 10920830 - -
          Affymetrix Rat Gene 1.1 ST Array 10914201 - -
          Affymetrix Rat Gene 1.1 ST Array 10920820 - -
          Rat Genome 230 2.0 Array 1387783_a_at - -
          Rat Genome 230 2.0 Array 1387783_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0800028730.rn.1 TC0800003538.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028731.rn.1 TC0800003538.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028732.rn.1 TC0800003538.rn.1 RTA TC
          Rat Expression Set 230 1387783_PM_a_at - -
          Rat Expression Set 230 1387783_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10914201 - -
          Affymetrix Rat Gene 1.0 ST Array 10920820 - -
          Rat Genome U34 Set J02749_at - -
          Rat Genome U34 Set J02749_g_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373487 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0