Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223897_g1
          See other RGD1559804 GE Assays ›
          Gene Symbol:
          RGD1559804
          Gene Name:
          similar to hypothetical protein 4930474N05
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 12191696 - 12194480 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04223897_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1559804
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113787.1 NP_001107259.1 4 1048 92
          GenBank mRNA BC158776.1 - 4 1048 7133601 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027799 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018361 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P027797 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074154 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027797 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027799 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074154 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027797 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P027799 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074154 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027797 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074154 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P027799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790529 - -
          Affymetrix Rat Gene 1.0 ST Array 10790528 - -
          Affymetrix Rat Gene 1.0 ST Array 10787045 - -
          Affymetrix Rat Gene 1.0 ST Array 10790530 - -
          Affymetrix Rat Gene 1.0 ST Array 10787047 - -
          Affymetrix Rat Gene 1.0 ST Array 10787044 - -
          Affymetrix Rat Gene 1.1 ST Array 10787043 - -
          Affymetrix Rat Gene 1.1 ST Array 10790527 - -
          Rat Genome 230 2.0 Array 1398021_PM_at - -
          Rat Genome 230 2.0 Array 1398021_at - -
          Affymetrix Rat Clariom S Assay PSR1600009616.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009614.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009605.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009607.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009609.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009610.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009606.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009615.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009617.rn.1 TC1600001255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009618.rn.1 TC1600001255.rn.1 RTA TC
          Rat Expression Set 230 1398021_at - -
          Rat Expression Set 230 1398021_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787043 - -
          Affymetrix Rat Gene 1.0 ST Array 10790527 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355105 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline