Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223964_g1
          See other RPL14 GE Assays ›
          Gene Symbol:
          Rpl14
          Gene Name:
          ribosomal protein L14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 129240528 - 129243400 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04223964_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022949.2 NP_075238.2 6 665 96
          GenBank mRNA BC168728.1 - 6 665 6918710 96
          FQ209993.1 - 3 412 96
          FQ210501.1 - 3 412 96
          FQ222022.1 - 3 412 96
          FQ222656.1 - 3 412 96
          FQ222698.1 - 3 412 96
          FQ223044.1 - 3 413 97
          FQ223231.1 - 3 96
          FQ223484.1 - 3 412 96
          FQ224076.1 - 3 412 96
          FQ225475.1 - 3 412 96
          FQ225874.1 - 3 412 96
          FQ229197.1 - 3 412 96
          FQ233060.1 - 3 412 96
          X94242.1 - 5 647 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl14

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          translation
          ribosomal large subunit biogenesis
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P412301 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035993 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035993 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P412301 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035993 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P412301 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P412301 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700144733 - -
          Agilent Rat cDNA Microarray (G4105A) 700144733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914367 - -
          Affymetrix Rat Gene 1.0 ST Array 10914370 - -
          Affymetrix Rat Gene 1.0 ST Array 10914368 - -
          Affymetrix Rat Gene 1.0 ST Array 10914369 - -
          Affymetrix Rat Gene 1.1 ST Array 10914366 - -
          Rat Genome 230 2.0 Array 1375375_PM_at - -
          Rat Genome 230 2.0 Array 1387887_PM_at - -
          Rat Genome 230 2.0 Array 1375375_at - -
          Rat Genome 230 2.0 Array 1387887_at - -
          Affymetrix HT Rat Focus Array 1375375_PM_at - -
          Affymetrix HT Rat Focus Array 1375375_at - -
          Affymetrix HT Rat Focus Array 1387887_at - -
          Affymetrix HT Rat Focus Array 1387887_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800014112.rn.1 TC0800001766.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800014111.rn.1 TC0800001766.rn.1 RTA TC
          Rat Expression Set 230 1375375_at - -
          Rat Expression Set 230 1375375_PM_at - -
          Rat Expression Set 230 1387887_at - -
          Rat Expression Set 230 1387887_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10914366 - -
          Rat Genome U34 Set X94242_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20869482 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351461 - -
          RatRef-12 Expression BeadChip ILMN_60945 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline