Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04224038_gH
          See other POLR2M GE Assays ›
          Gene Symbol:
          Polr2m
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide M
          Gene Aliases:
          Glurr, Grinl1a
          Chromosome Location:
          Chr.8: 77985190 - 77992621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04224038_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2m
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183402.1 NP_899652.1 4 1244 91
          GenBank mRNA AF326772.2 - 4 1244 91
          AY724533.1 - 4 1345 91
          BC079379.1 - 4 1312 7110855 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Polr2m

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA biosynthetic process
          maintenance of ER location
          DNA-directed RNA polymerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P077892 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032158 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033909 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012178 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112153 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077892 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032158 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077892 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032158 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032158 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012178 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P077892 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205301 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P347938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918581 - -
          Affymetrix Rat Gene 1.0 ST Array 10809498 - -
          Affymetrix Rat Gene 1.0 ST Array 10771166 - -
          Affymetrix Rat Gene 1.0 ST Array 10918582 - -
          Affymetrix Rat Gene 1.0 ST Array 10771168 - -
          Affymetrix Rat Gene 1.0 ST Array 10918584 - -
          Affymetrix Rat Gene 1.0 ST Array 10771167 - -
          Affymetrix Rat Gene 1.0 ST Array 10771165 - -
          Affymetrix Rat Gene 1.0 ST Array 10918583 - -
          Affymetrix Rat Gene 1.0 ST Array 10809499 - -
          Affymetrix Rat Gene 1.0 ST Array 10918585 - -
          Affymetrix Rat Gene 1.1 ST Array 10918580 - -
          Affymetrix Rat Gene 1.1 ST Array 10809497 - -
          Affymetrix Rat Gene 1.1 ST Array 10771134 - -
          Rat Genome 230 2.0 Array 1371051_at - -
          Rat Genome 230 2.0 Array 1371051_PM_at - -
          Rat Genome 230 2.0 Array 1398889_PM_at - -
          Rat Genome 230 2.0 Array 1398889_at - -
          Affymetrix HT Rat Focus Array 1398889_at - -
          Affymetrix HT Rat Focus Array 1371051_at - -
          Affymetrix HT Rat Focus Array 1371051_PM_at - -
          Affymetrix HT Rat Focus Array 1398889_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800023269.rn.1 TC0800002878.rn.1 RTA TC
          Rat Expression Set 230 1371051_at - -
          Rat Expression Set 230 1398889_PM_at - -
          Rat Expression Set 230 1371051_PM_at - -
          Rat Expression Set 230 1398889_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918580 - -
          Rat Genome U34 Set rc_AI229689_at - -
          Rat Genome U34 Set rc_AI171953_at - -
          Rat Genome U34 Set H34023_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20855809 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365030 - -
          RatRef-12 Expression BeadChip ILMN_62918 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline