Search
Search
| Gene Symbol: Pja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101006.1 | NP_001094476.1 | 3 | 653 | 76 | |
| XM_006257105.2 | XP_006257167.1 | 2 | 1767 | 76 | ||
| XM_006257106.2 | XP_006257168.1 | 2 | 1484 | 76 | ||
| XM_006257107.2 | XP_006257169.1 | 3 | 1227 | 76 | ||
| GenBank mRNA | BC081885.1 | - | 3 | 653 | 7108525 | 76 |
Set Membership: |
|