Search
Search

| Gene Symbol: Mir425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032277.1 | 1 | 41 | 74 | ||
| Gene Symbol: Ndufaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243845.2 | XP_006243907.1 | 1 | 74 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3555 | Mir3555 |
Mir191b, rno-mir-191b, rno-mir-3555 |
-354.0 | Anti-sense | Chr. 8 - 117354348 - 117354467 on NCBI Build Rnor_6.0 |
| Downstream | DALR anticodon binding domain containing 3 | Dalrd3 | 1.0kb | Sense | Chr. 8 - 117355943 - 117358819 on NCBI Build Rnor_6.0 |
Set Membership: |
|