Search
Search
| Gene Symbol: Zbtb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191670.1 | NP_001178599.1 | 7 - 8 | 1872 | 70 | |
| XM_006249937.2 | XP_006249999.1 | 8 - 9 | 2101 | 70 | ||
| XM_006249938.2 | XP_006250000.1 | 8 - 9 | 2066 | 70 | ||
| XM_006249939.2 | XP_006250001.1 | 8 - 9 | 2092 | 70 | ||
| XM_006249940.2 | XP_006250002.1 | 8 - 9 | 2144 | 70 | ||
Set Membership: |
|