Search
Search

Tsga10ip| Gene Symbol: Tsga10ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004278.3 | NP_001004278.2 | 3 - 4 | 1006 | 63 | |
| NM_001270736.1 | NP_001257665.1 | 3 - 4 | 1006 | 63 | ||
| NM_001270737.1 | NP_001257666.1 | 3 - 4 | 1006 | 63 | ||
| XM_006230847.2 | XP_006230909.1 | 3 - 4 | 1281 | 63 | ||
| XM_008760155.1 | XP_008758377.1 | 3 - 4 | 1291 | 63 | ||
| XM_008760156.1 | XP_008758378.1 | 3 - 4 | 1290 | 63 | ||
| XM_008760157.1 | XP_008758379.1 | 3 - 4 | 1292 | 63 | ||
| GenBank mRNA | AY690667.1 | - | 3 - 4 | 1023 | 63 | |
| BC097254.1 | - | 3 - 4 | 987 | 7453704 | 63 | |
Set Membership: |
|