Search
Search
| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017464.2 | NP_001017464.1 | 7 - 8 | 835 | 62 | |
| XM_006230403.2 | XP_006230465.1 | 7 - 8 | 935 | 62 | ||
| XM_008759903.1 | XP_008758125.1 | 7 - 8 | 935 | 62 | ||
| XM_008759904.1 | XP_008758126.1 | 5 - 6 | 601 | 62 | ||
| XM_008759905.1 | XP_008758127.1 | 7 - 8 | 935 | 62 | ||
| XM_008759906.1 | XP_008758128.1 | 7 - 8 | 935 | 62 | ||
| GenBank mRNA | BC079317.1 | - | 7 - 8 | 838 | 7135047 | 62 |
Set Membership: |
|