Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04337847_m1
          See other CRISP4 GE Assays ›
          Gene Symbol:
          Crisp4
          Gene Name:
          cysteine-rich secretory protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 24356577 - 24383679 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04337847_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crisp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039393.2 NP_001034482.2 7 - 8 799 117
          XM_008766935.1 XP_008765157.1 7 - 8 799 117
          GenBank mRNA DQ200907.1 - 7 - 8 799 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001682.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protease 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          binding of sperm to zona pellucida
          regulation of acrosome reaction
          calcium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121474 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62570 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926833 - -
          Affymetrix Rat Gene 1.0 ST Array 10926835 - -
          Affymetrix Rat Gene 1.0 ST Array 10926827 - -
          Affymetrix Rat Gene 1.0 ST Array 10926828 - -
          Affymetrix Rat Gene 1.0 ST Array 10926826 - -
          Affymetrix Rat Gene 1.0 ST Array 10926836 - -
          Affymetrix Rat Gene 1.0 ST Array 10926824 - -
          Affymetrix Rat Gene 1.0 ST Array 10926834 - -
          Affymetrix Rat Gene 1.0 ST Array 10926825 - -
          Affymetrix Rat Gene 1.1 ST Array 10926823 - -
          Affymetrix Rat Clariom S Assay PSR0900013784.rn.1 TC0900001682.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10926823 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22227451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline