Search
Search
| Gene Symbol: Rtdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127557.2 | NP_001121029.1 | 1 - 2 | 243 | 71 | |
| XM_006256340.2 | XP_006256402.1 | 2 - 3 | 299 | 71 | ||
| XM_006256341.2 | XP_006256403.1 | 2 - 3 | 352 | 71 | ||
| GenBank mRNA | BC162057.1 | - | 1 - 2 | 245 | 7114691 | 71 |
Set Membership: |
|