Search
Search
| Gene Symbol: Nr1i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270838.1 | NP_001257767.1 | 3 - 4 | 423 | 83 | |
| NM_001270839.1 | NP_001257768.1 | 3 - 4 | 423 | 83 | ||
| NM_001270840.1 | NP_001257769.1 | 3 - 4 | 423 | 83 | ||
| NM_022941.4 | NP_075230.1 | 3 - 4 | 423 | 83 | ||
| XM_006250261.2 | XP_006250323.1 | 3 - 4 | 532 | 83 | ||
| XM_008769758.1 | XP_008767980.1 | 3 - 4 | 535 | 83 | ||
| XM_008769759.1 | XP_008767981.1 | 3 - 4 | 535 | 83 | ||
| GenBank mRNA | AB104736.1 | - | 3 - 4 | 476 | 83 | |
| AB105072.1 | - | 3 - 4 | 476 | 83 | ||
| AB204897.1 | - | 2 - 3 | 274 | 83 | ||
| AB204898.1 | - | 2 - 3 | 274 | 83 | ||
| AB204899.1 | - | 2 - 3 | 274 | 83 | ||
| AB204900.1 | - | 2 - 3 | 274 | 83 | ||
| AF133094.1 | - | 2 - 3 | 274 | 83 | ||
| AF133095.1 | - | 2 - 3 | 274 | 83 | ||
| BC089763.1 | - | 3 - 4 | 423 | 7324318 | 83 | |
Set Membership: |
|