Search
Search

Tnfrsf11a| Gene Symbol: Tnfrsf11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271235.1 | NP_001258164.1 | 9 - 10 | 1566 | 63 | |
| XM_008769487.1 | XP_008767709.1 | 9 - 10 | 1544 | 63 | ||
Set Membership: |
|