Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04932425_m1
          See other GALR1 GE Assays ›
          Gene Symbol:
          Galr1
          Gene Name:
          galanin receptor 1
          Gene Aliases:
          Galnr1
          Chromosome Location:
          Chr.18: 79243009 - 79258570 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04932425_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012958.2 NP_037090.2 1 - 2 892 99
          GenBank mRNA U30290.1 - 1 - 2 906 99

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galr1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of cortisol secretion
          galanin receptor activity
          peptide hormone binding
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11633 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116100 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75222 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805467 - -
          Affymetrix Rat Gene 1.0 ST Array 10805466 - -
          Affymetrix Rat Gene 1.1 ST Array 10805465 - -
          Rat Genome 230 2.0 Array 1387702_PM_at - -
          Rat Genome 230 2.0 Array 1387702_at - -
          Affymetrix HT Rat Focus Array 1387702_at - -
          Affymetrix HT Rat Focus Array 1387702_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800012553.rn.1 TC1800001798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800012552.rn.1 TC1800001798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800012550.rn.1 TC1800001798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800012551.rn.1 TC1800001798.rn.1 RTA TC
          Rat Expression Set 230 1387702_PM_at - -
          Rat Expression Set 230 1387702_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805465 - -
          Rat Genome U34 Set U30290_at - -
          Rat Genome U34 Set U30290_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356924 - -
          RatRef-12 Expression BeadChip ILMN_67613 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline