Search
Search

Supt4h1| Gene Symbol: Supt4h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105828.2 | NP_001099298.2 | 5 | 335 | 61 | |
| GenBank mRNA | BC168730.1 | - | 5 | 335 | 6887426 | 61 |
| FQ213593.1 | - | 5 | 342 | 61 | ||
| FQ217757.1 | - | 5 | 342 | 61 | ||
| FQ224188.1 | - | 5 | 331 | 61 | ||
| FQ228457.1 | - | 5 | 329 | 61 | ||
Set Membership: |
|