Search
Search

RGD1308112| Gene Symbol: RGD1308112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249895.2 | XP_006249957.1 | 3 - 4 | 556 | 77 | |
| XM_008769575.1 | XP_008767797.1 | 2 - 3 | 473 | 77 | ||
| XM_008769576.1 | XP_008767798.1 | 3 - 4 | 551 | 77 | ||
Set Membership: |
|