Search
Search

RGD1560986| Gene Symbol: RGD1560986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759244.1 | XP_008757466.1 | 5 - 6 | 1008 | 60 | |
| XM_008759245.1 | XP_008757467.1 | 5 - 6 | 1008 | 60 | ||
| XM_008759246.1 | XP_008757468.1 | 6 - 7 | 933 | 60 | ||
| XM_008759247.1 | XP_008757469.1 | 6 - 7 | 933 | 60 | ||
| XM_008759248.1 | XP_008757470.1 | 7 - 8 | 858 | 60 | ||
| XM_008759249.1 | XP_008757471.1 | 5 - 6 | 1008 | 60 | ||
| XM_008759250.1 | XP_008757472.1 | 5 - 6 | 1008 | 60 | ||
| XM_008759251.1 | XP_008757473.1 | 5 - 6 | 1008 | 60 | ||
| XM_008759252.1 | XP_008757474.1 | 5 - 6 | 1008 | 60 | ||
Set Membership: |
|