Search
Search

RGD1562063| Gene Symbol: RGD1562063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241913.2 | XP_006241975.1 | 6 - 7 | 1970 | 58 | |
| XM_006241914.2 | XP_006241976.1 | 6 - 7 | 1970 | 58 | ||
| XM_008765632.1 | XP_008763854.1 | 6 - 7 | 1970 | 58 | ||
Set Membership: |
|